Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7438G-100G PC7438G-25G PC7438G-500G
77417-890EA 74.38 USD InStock
77417-890 77417-892 77417-894
4-(Trifluoromethoxy)benzonitrile 98%
4-(Trifluoromethoxy)benzonitrile


Formula: C₈H₄F₃NO
MW: 187.12
Boiling Pt: 192…193 °C
Density: 1.284
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00039474
CAS Number: 332-25-2
EINECS: 206-363-3
UN: 3276
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty