Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluoropyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7951-100G PC7951-25G PC7951-5G
77418-318EA 158.94 USD InStock
77418-318 77418-320 77418-322
3-Bromo-2-fluoropyridine 98%
3-Bromo-2-fluoropyridine


Formula: C₅H₃BrFN
MW: 175.99
Boiling Pt: 76 °C/20 mmHg
Flash Pt: <90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112496
CAS Number: 36178-05-9
UN: 2810
ADR: 6.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty