Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxyindole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0581-10G OR0581-1G OR0581-25G
77402-146EA 49.92 USD InStock
77402-146 77402-148 77402-150
6-Methoxyindole 98%
6-Methoxyindole


Formula: C₉H₉NO
Boiling Pt: 105 °C/0.2 mmHg
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00022780
CAS Number: 3189-13-7
EINECS: 221-689-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty