Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Erbium trichloride, anhydrous 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

IN1695-100G IN1695-25G
77401-432EA 363.72 USD InStock
77401-432 77401-434
Erbium trichloride, anhydrous 99,9%
Erbium trichloride


Formula: Cl₃Er
MW: 273.62
Boiling Pt: 1500
Melting Pt: 751
Density: 4.1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010989
CAS Number: 10138-41-7
EINECS: 233-385-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty