Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-2-Amino-1-propanol

Supplier: APOLLO SCIENTIFIC
Danger

OR0903-100G OR0903-25G
77402-292EA 166.06 USD InStock
77402-292 77402-294
(R)-(-)-2-Amino-1-propanol
(R)-(-)-2-Amino-1-propanol
Chiral intermediate for pharmaceuticals and agrochemicals.


Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 173
Density: 0.965 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00064413
CAS Number: 35320-23-1
UN: 2735
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty