Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Amino-2,2,2-trifluoroethane 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC7320-25G PC7320-5G
77417-814EA 72.34 USD InStock
77417-814 77417-816
1-Amino-2,2,2-trifluoroethane 99%
1-Amino-2,2,2-trifluoroethane


Formula: C₂H₄F₃N
Boiling Pt: 36…38 °C
Density: 1.259 g/cm³ (25 °C)
Flash Pt: –16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008132
CAS Number: 753-90-2
EINECS: 212-041-3
UN: 3286
ADR: 3+6.1+8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty