Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω,3-Dibromoacetophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR5960-100G OR5960-25G OR5960-5G
77410-608EA 399.38 USD InStock
77410-608 77410-610 77410-612
ω,3-Dibromoacetophenone 97%
ω,3-Dibromoacetophenone


Formula: C₈H₆Br₂O
Melting Pt: 47…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00082698
CAS Number: 18523-22-3
UN: 3261
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty