Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,3,4-Trifluorophenyl)acetonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7818J-1G PC7818J-25G PC7818J-5G
77418-222EA 21.28 USD InStock
77418-222 77418-224 77418-226
(2,3,4-Trifluorophenyl)acetonitrile 97%
(2,3,4-Trifluorophenyl)acetonitrile


Formula: C₈H₄F₃N
Storage Temperature: Ambient
MDL Number: MFCD00061234
CAS Number: 243666-13-9
UN: 3276
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty