Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-(trifluoromethyl)benzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4374Q-25G PC4374Q-5G
77415-306EA 268.96 USD InStock
77415-306 77415-308
3-Fluoro-5-(trifluoromethyl)benzoyl chloride 97%
3-Fluoro-5-(trifluoromethyl)benzoyl chloride


Formula: C₈H₃ClF₄O
MW: 226.56
Boiling Pt: 119
Density: 1.482 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00061157
CAS Number: 171243-30-4
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty