Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethylbenzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR24121-100MG OR24121-250MG
77407-422EA 21.36 USD InStock
77407-422 77407-424
2,4-Dimethylbenzoyl chloride
2,4-Dimethylbenzoyl chloride


Formula: C₉H₉ClO
Boiling Pt: 82 °C/1 mmHg
Melting Pt: 24…26 °C
Storage Temperature: Ambient
MDL Number: MFCD00798005
CAS Number: 21900-42-5
UN: 3265
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty