Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Triisopropylbenzenesulphonyl chloride 98+%

Supplier: APOLLO SCIENTIFIC
Danger

OR5360-100G OR5360-25G
77410-308EA 55.02 USD InStock
77410-308 77410-310
2,4,6-Triisopropylbenzenesulphonyl chloride 98+%
2,4,6-Triisopropylbenzenesulphonyl chloride


Formula: C₁₅H₂₃ClO₂S
MW: 302.87
Melting Pt: 94…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00007433
CAS Number: 6553-96-4
EINECS: 229-479-6
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty