Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-(trifluoromethoxy)aniline 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC1593K-100G PC1593K-25G PC1593K-500G
77413-234EA 65.84 USD InStock
77413-234 77413-236 77413-238
4-Bromo-2-(trifluoromethoxy)aniline 99%
4-Bromo-2-(trifluoromethoxy)aniline


Formula: C₇H₅BrF₃NO
Boiling Pt: 98…100 °C/15 mmHg
Melting Pt: 29…30 °C
Density: 1.691 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00179338
CAS Number: 175278-09-8
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty