Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium hexafluoroantimonate 99%

Supplier: APOLLO SCIENTIFIC

PC6279-100G PC6279-25G PC6279-5G
77417-350EA 312.78 USD InStock
77417-350 77417-352 77417-354
Potassium hexafluoroantimonate 99%
Potassium hexafluoroantimonate


Formula: F₆KSb
MW: 274.85
Boiling Pt: 1505 °C
Melting Pt: 846 °C
Storage Temperature: Ambient
MDL Number: MFCD00043080
CAS Number: 16893-92-8
UN: 1549
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty