Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)benzylamine 94%

Supplier: APOLLO SCIENTIFIC
Danger

PC7630-100G PC7630-25G
77418-040EA 115.12 USD InStock
77418-040 77418-042
4-(Trifluoromethyl)benzylamine 94%
4-(Trifluoromethyl)benzylamine


Formula: C₈H₈F₃N
Boiling Pt: 80…82 °C/15 mmHg
Density: 1.229 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00010220
CAS Number: 3300-51-4
EINECS: 221-971-9
UN: 2735
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty