Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Bis(trifluoromethyl)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC5969-1G PC5969-25G PC5969-5G
77417-216EA 21.36 USD InStock
77417-216 77417-218 77417-220
2,5-Bis(trifluoromethyl)benzyl bromide
2,5-Bis(trifluoromethyl)benzyl bromide


Formula: C₉H₅BrF₆
MW: 307.03
Boiling Pt: 176 °C
Density: 1.674 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00799485
CAS Number: 302911-98-4
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty