Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-(trifluoromethyl)benzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC4376E-25G PC4376E-5G PC4376E-1G
77415-334EA 322.96 USD InStock
77415-334 77415-336 77415-332
2-Fluoro-3-(trifluoromethyl)benzyl bromide 98%
2-Fluoro-3-(trifluoromethyl)benzyl bromide


Formula: C₈H₅BrF₄
MW: 257.03
Boiling Pt: 197
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00061172
CAS Number: 184970-25-0
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty