Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-4-fluorobenzotrifluoride 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2403-1G PC2403-25G PC2403-5G
77413-628EA 62.16 USD InStock
77413-628 77413-630 77413-632
3,5-Dichloro-4-fluorobenzotrifluoride 98%
3,5-Dichloro-4-fluorobenzotrifluoride


Formula: C₇H₂Cl₂F₄
MW: 232.99 g/mol
Boiling Pt: 166…168 °C
Density: 1.555 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00068183
CAS Number: 77227-81-7
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty