Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-o-anisidine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC5085-1G PC5085-25G PC5085-5G
77416-416EA 21.32 USD InStock
77416-416 77416-418 77416-420
3-Fluoro-o-anisidine 97%
3-Fluoro-o-anisidine


Formula: C₇H₈FNO
MW: 141.15
Boiling Pt: 94 °C/10 mmHg
Density: 1.172 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00007759
CAS Number: 437-83-2
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty