Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-o-phenylenediamine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR2703-100G OR2703-25G OR2703-500G
77407-880EA 74.38 USD InStock
77407-880 77407-882 77407-884
4-Chloro-o-phenylenediamine 97%
4-Chloro-o-phenylenediamine


Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Boiling Pt: 280
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00011691
CAS Number: 95-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty