Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-3-fluoro-4-iodobenzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC0053-100G PC0053-25G PC0053-500G
77412-360EA 62.64 USD InStock
77412-360 77412-362 77412-364
1-Chloro-3-fluoro-4-iodobenzene 97%
1-Chloro-3-fluoro-4-iodobenzene


Formula: C₆H₃ClFI
MW: 256.45 g/mol
Boiling Pt: 107 °C/18 mmHg
Melting Pt: 17…19 °C
Storage Temperature: Ambient
MDL Number: MFCD00079729
CAS Number: 6797-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty