Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenylbenzophenone 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR18324-100G OR18324-500G
77406-228EA 34.26 USD InStock
77406-228 77406-230
4-Phenylbenzophenone 99%
4-Phenylbenzophenone


Formula: C₁₉H₁₄O
MW: 258.32 g/mol
Boiling Pt: 419
Melting Pt: 98…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00003079
CAS Number: 2128-93-0
EINECS: 218-345-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty