Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethyl)uracil 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7780-25G PC7780-100G
77418-194EA 244.02 USD InStock
77418-194 77418-190 77418-192
5-(Trifluoromethyl)uracil 98%
5-(Trifluoromethyl)uracil


Formula: C₅H₃F₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00006024
CAS Number: 54-20-6
EINECS: 200-197-5
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty