Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,7-Dibromonaphthalene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR19537-10G OR19537-5G
77406-486EA 58.08 USD InStock
77406-486 77406-490
2,7-Dibromonaphthalene 98%
2,7-Dibromonaphthalene


Formula: C₁₀H₆Br₂
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00238580
CAS Number: 58556-75-5
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty