Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Aminonicotinonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR6119-100G OR6119-25G OR6119-5G
77410-708EA 528.76 USD InStock
77410-708 77410-710 77410-712
6-Aminonicotinonitrile 97%
6-Aminonicotinonitrile


Formula: C₆H₅N₃
Boiling Pt: 240…250 °C/15 mmHg
Melting Pt: 159…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00128902
CAS Number: 4214-73-7
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty