Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetyl-3-chlorothiophene

Supplier: APOLLO SCIENTIFIC
Danger

OR15400-1G OR15400-5G
77405-394EA 44.82 USD InStock
77405-394 77405-396
2-Acetyl-3-chlorothiophene
2-Acetyl-3-chlorothiophene


Formula: C₆H₅ClOS
MW: 160.62 g/mol
Boiling Pt: 117…118 °C/15 mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00794248
CAS Number: 89581-82-8
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty