Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Chloromethyl)uracil

Supplier: APOLLO SCIENTIFIC
Warning

OR1213-100MG OR1213-250MG
77404-196EA 21.34 USD InStock
77404-196 77404-198
5-(Chloromethyl)uracil
5-(Chloromethyl)uracil


Formula: C₅H₅ClN₂O₂
Melting Pt: >350 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00218445
CAS Number: 3590-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty