Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Nitro-1H-indazole 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR13550-25G OR13550-5G
77404-662EA 96.78 USD InStock
77404-662 77404-664
7-Nitro-1H-indazole 97%
7-Nitro-1H-indazole


Formula: C₇H₅N₃O₂
Storage Temperature: Freezer
MDL Number: MFCD00022789
CAS Number: 2942-42-9
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty