Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,O-Bis(trimethylsilyl)acetamide 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR5155A-100G OR5155A-25G OR5155A-500G
77410-154EA 28.52 USD InStock
77410-154 77410-156 77410-158
N,O-Bis(trimethylsilyl)acetamide 97%
N,O-Bis(trimethylsilyl)acetamide


Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Boiling Pt: 162
Melting Pt: 24
Density: 0.832 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008270
CAS Number: 10416-59-8
EINECS: 233-892-7
UN: 2920
ADR: 8+3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty