Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridinium chloride 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR13636-100G OR13636-25G
77404-690EA 34.64 USD InStock
77404-690 77404-692
Pyridinium chloride 95%
Pyridinium chloride


Formula: C₅H₆ClN
MW: 115.56 g/mol
Boiling Pt: 222.5
Melting Pt: 144
Storage Temperature: Ambient
MDL Number: MFCD00012802
CAS Number: 628-13-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty