Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylphenylboronic acid 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR10312-25G OR10312-5G OR10312-1G
77402-580EA 497.18 USD InStock
77402-580 77402-582 77402-578
2-Acetylphenylboronic acid 95+%
2-Acetylphenylboronic acid


Formula: C₈H₉BO₃
Melting Pt: 170 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD01321263
CAS Number: 308103-40-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty