Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α'-Chloro-α,α,α-trifluoro-p-xylene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7633G-25G PC7633G-5G
77418-066EA 36.68 USD InStock
77418-066 77418-068
α'-Chloro-α,α,α-trifluoro-p-xylene 98%
α'-Chloro-α,α,α-trifluoro-p-xylene


Formula: C₈H₆ClF₃
MW: 194.58
Boiling Pt: 68 °C/12 mmHg
Melting Pt: 19
Density: 1.315 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00040772
CAS Number: 939-99-1
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty