Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (methylthio)acetate

Supplier: APOLLO SCIENTIFIC
Warning

OR9387-100G OR9387-25G OR9387-5G
77412-046EA 261.84 USD InStock
77412-046 77412-048 77412-050
Methyl (methylthio)acetate
Methyl (methylthio)acetate


Formula: C₄H₈O₂S
Boiling Pt: 49…51 °C/12 mmHg
Density: 1.11 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039809
CAS Number: 16630-66-3
EINECS: 240-683-4
UN: 3272
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty