Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-(trifluoromethyl)pyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC1264-100G PC1264-25G PC1264-5G
77413-116EA 172.18 USD InStock
77413-116 77413-118 77413-120
2-Fluoro-6-(trifluoromethyl)pyridine 97%
2-Fluoro-6-(trifluoromethyl)pyridine


Formula: C₆H₃F₄N
Boiling Pt: 143 °C
Melting Pt: –15.2 °C
Density: 1.4 g/cm³ (25 °C)
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03092901
CAS Number: 94239-04-0
EINECS: 428-100-3
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty