Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(trifluoromethyl)benzenesulphonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC3085-25G PC3085-5G
77414-450EA 88.64 USD InStock
77414-450 77414-452
3,5-Bis(trifluoromethyl)benzenesulphonyl chloride
3,5-Bis(trifluoromethyl)benzenesulphonyl chloride


Formula: C₈H₃ClF₆O₂S
MW: 312.62
Melting Pt: 34…38 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00014725
CAS Number: 39234-86-1
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty