Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-2-nitro-6-picoline 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR16078-100G OR16078-25G OR16078-5G
77405-628EA 611.28 USD InStock
77405-628 77405-630 77405-632
3-Hydroxy-2-nitro-6-picoline 95%
3-Hydroxy-2-nitro-6-picoline


Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00006259
CAS Number: 15128-90-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty