Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenoxybenzenesulphonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR12614-100MG OR12614-1G OR12614-250MG
77404-382EA 21.4 USD InStock
77404-382 77404-384 77404-386
4-Phenoxybenzenesulphonyl chloride
4-Phenoxybenzenesulphonyl chloride


Formula: C₁₂H₉ClO₃S
MW: 268.72
Boiling Pt: 180 °C/5 mmHg
Melting Pt: 44
Storage Temperature: Ambient
MDL Number: MFCD00625748
CAS Number: 1623-92-3
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty