Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-chloro-3-pyridineboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR2583-1G OR2583-250MG OR2583-5G
77407-668EA 194.6 USD InStock
77407-668 77407-670 77407-672
5-Bromo-2-chloro-3-pyridineboronic acid 97%
5-Bromo-2-chloro-3-pyridineboronic acid


Formula: C₅H₄BBrClNO₂
MW: 236.26
Melting Pt: 148…150 °C
Storage Temperature: Freezer
MDL Number: MFCD09037486
CAS Number: 1072944-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty