Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl fluoromalonate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC3013-25G PC3013-5G
77414-090EA 24.46 USD InStock
77414-090 77414-092
Dimethyl fluoromalonate 97%
Dimethyl fluoromalonate
Active fluoromethylene group, useful reagent for the synthesis of fluoro-heterocycles, e.g Pyrimidines; also can produce fluorinated trisubstituted alkenes, ask for refs.


Formula: C₅H₇FO₄
MW: 150.11 g/mol
Boiling Pt: 111…112 °C/45 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00054680
CAS Number: 344-14-9
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty