Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-4-Fluoro-α-methylbenzylamine 96%

Supplier: APOLLO SCIENTIFIC
Danger

PC3823CM-25G PC3823CM-50G PC3823CM-5G
77414-940EA 75.4 USD InStock
77414-940 77414-942 77414-944
(±)-4-Fluoro-α-methylbenzylamine 96%
(±)-4-Fluoro-α-methylbenzylamine


Formula: C₈H₁₀FN
MW: 139.17
Boiling Pt: 76 °C/22 mmHg
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00041323
CAS Number: 403-40-7
EINECS: 206-958-8
UN: 2735
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty