Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Diethylamino)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC

OR900360-100G OR900360-25G OR900360-5G
77411-490EA 613.32 USD InStock
77411-490 77411-492 77411-494
4-(Diethylamino)benzonitrile 98%
4-(Diethylamino)benzonitrile


Formula: C₁₁H₁₄N₂
Storage Temperature: Ambient
MDL Number: MFCD00019754
CAS Number: 2873-90-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty