Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetamido-5-aminopyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR4098-25G OR4098-5G
77409-600EA 405.48 USD InStock
77409-600 77409-602
2-Acetamido-5-aminopyridine 98%
2-Acetamido-5-aminopyridine


Formula: C₇H₉N₃O
Melting Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00160311
CAS Number: 29958-14-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty