Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromopicolinic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR13059-25G OR13059-5G
77404-504EA 47.88 USD InStock
77404-504 77404-506
5-Bromopicolinic acid 97%
5-Bromopicolinic acid


Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00234149
CAS Number: 30766-11-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty