Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentaethylene glycol 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR13897-10G OR13897-25G OR13897-5G
77404-762EA 75.4 USD InStock
77404-762 77404-764 77404-766
Pentaethylene glycol 97%
Pentaethylene glycol


Formula: C₁₀H₂₂O₆
Boiling Pt: 338…340 °C
Density: 1.126 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty