Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl acetylenedicarboxylate 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR59420-100G OR59420-25G
77410-588EA 74.38 USD InStock
77410-588 77410-590
Diethyl acetylenedicarboxylate 95%
Diethyl acetylenedicarboxylate


Formula: C₈H₁₀O₄
MW: 170.16 g/mol
Boiling Pt: 107…110 °C/11 mmHg
Melting Pt: 1…2 °C
Density: 1.063 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009186
CAS Number: 762-21-0
EINECS: 212-095-8
UN: 3265
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty