Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromobenzo[b]thiophene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR23397-25G OR23397-5G
77407-318EA 245.54 USD InStock
77407-318 77407-320
5-Bromobenzo[b]thiophene 98%
5-Bromobenzo[b]thiophene


Formula: C₈H₅BrS
Boiling Pt: 105…107 °C/10 mmHg
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD03069318
CAS Number: 4923-87-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty