Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylthiophene-2-carboxaldehyde 90%

Supplier: APOLLO SCIENTIFIC
Warning

OR10155-100G OR10155-25G
77402-506EA 93.74 USD InStock
77402-506 77402-508
3-Methylthiophene-2-carboxaldehyde 90%
3-Methylthiophene-2-carboxaldehyde


Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 215 °C
Density: 1.17 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005430
CAS Number: 5834-16-2
EINECS: 227-418-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty