Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitro-4-(trifluoromethyl)phenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC

PC8009-10G PC8009-1G PC8009-5G
77418-328EA 149.76 USD InStock
77418-328 77418-330 77418-332
2-Nitro-4-(trifluoromethyl)phenylacetonitrile 98%
2-Nitro-4-(trifluoromethyl)phenylacetonitrile


Formula: C₉H₅F₃N₂O₂
Boiling Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00045621
CAS Number: 13544-06-4
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty