Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Tetrahydrofurfuryl bromide 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR23265-100G OR23265-500G
77407-200EA 154.86 USD InStock
77407-200 77407-202
(±)-Tetrahydrofurfuryl bromide 95%
(±)-Tetrahydrofurfuryl bromide


Formula: C₅H₉BrO
Boiling Pt: 170…171 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016894
CAS Number: 1192-30-9
EINECS: 214-750-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty