Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Bromomethyl)tetrahydropyran 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR10382-1G OR10382-25G OR10382-5G
77402-662EA 21.26 USD InStock
77402-662 77402-664 77402-666
4-(Bromomethyl)tetrahydropyran 97%
4-(Bromomethyl)tetrahydropyran


Formula: C₆H₁₁BrO
MW: 179.0549 g/mol
Boiling Pt: 89…93 °C/10 mmHg
Storage Temperature: Freezer
MDL Number: MFCD03788561
CAS Number: 125552-89-8
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty