Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Diphenylhydrazine hydrochloride 98%

Supplier: APOLLO SCIENTIFIC

OR911932-25G OR911932-5G
77411-634EA 106.4 USD InStock
77411-634 77411-636 77411-632
1,1-Diphenylhydrazine hydrochloride 98%
1,1-Diphenylhydrazine hydrochloride


Formula: C₁₂H₁₃ClN₂
Melting Pt: 162…166 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012950
CAS Number: 530-47-2
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty